Assay Details
| Gene Symbol: Gnai3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010306.3 | NP_034436.1 | 5 - 6 | 728 | 88 | |
| GenBank mRNA | AK004566.1 | - | 5 - 6 | 722 | 88 | |
| AK077490.1 | - | 5 - 6 | 690 | 88 | ||
| AK133749.1 | - | 5 - 6 | 669 | 88 | ||
| AK167438.1 | - | 5 - 6 | 682 | 88 | ||
| AK168578.1 | - | 5 - 6 | 666 | 88 | ||
| AK169067.1 | - | 5 - 6 | 673 | 88 | ||
| AK169285.1 | - | 5 - 6 | 703 | 88 | ||
| BC003462.1 | - | 5 - 6 | 654 | 88 | ||
| BC020009.1 | - | 5 - 6 | 700 | 88 | ||
| BC041107.1 | - | 5 - 6 | 639 | 2648164 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gnai3
Genomic Map