Assay Details
| Gene Symbol: Denr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026603.4 | NP_080879.1 | 1 - 2 | 95 | 100 | |
| GenBank mRNA | AK010394.1 | - | 1 - 2 | 112 | 100 | |
| AK017647.1 | - | 1 - 2 | 124 | 100 | ||
| AK018762.1 | - | 1 - 2 | 96 | 100 | ||
| AK135127.1 | - | 1 - 2 | 131 | 100 | ||
| BC025143.1 | - | 1 - 2 | 79 | 100 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK017647.1, AK018762.1, AK135127.1, NM_026603.4 transcripts.
Set Membership: |
|

Denr
Genomic Map