Assay Details
| Gene Symbol: Abcb9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019875.2 | NP_063928.2 | 9 - 10 | 1966 | 76 | |
| GenBank mRNA | AB045382.2 | - | 8 - 9 | 1786 | 76 | |
| AB093298.1 | - | 9 - 10 | 1948 | 76 | ||
| AF216495.1 | - | 9 - 10 | 1959 | 76 | ||
| AK020749.1 | - | 3 - 4 | 666 | 76 | ||
| AK044140.1 | - | 9 - 10 | 1944 | 76 | ||
| BC053014.1 | - | 9 - 10 | 1919 | 5686698 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Abcb9
Genomic Map