Assay Details
| Gene Symbol: Pitpnm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289472.1 | NP_001276401.1 | 19 - 20 | 3294 | 77 | |
| NM_011256.3 | NP_035386.1 | 19 - 20 | 3323 | 77 | ||
| GenBank mRNA | AF058693.1 | - | 18 - 19 | 2991 | 77 | |
| AK129364.1 | - | 19 - 20 | 3294 | 77 | ||
| AK149575.1 | - | 2 - 3 | 731 | 77 | ||
| BC070452.1 | - | 16 - 17 | 2583 | 30621943 | 77 | |
| BC082303.1 | - | 19 - 20 | 3323 | 77 | ||
More Information
Set Membership: |
|

Pitpnm2
Genomic Map