Assay Details
| Gene Symbol: Mybphl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026831.1 | NP_081107.1 | 2 - 3 | 324 | 95 | |
| GenBank mRNA | AK004148.1 | - | 2 - 3 | 325 | 95 | |
| BC089625.1 | - | 2 - 3 | 312 | 6756382 | 95 | |
| BC132636.1 | - | 2 - 3 | 313 | 40130913 | 95 | |
| BC132638.1 | - | 2 - 3 | 313 | 40130915 | 95 | |
| BY704393.1 | - | 2 - 3 | 325 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mybphl
Genomic Map