Assay Details
| Gene Symbol: Mybphl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026831.1 | NP_081107.1 | 5 - 6 | 818 | 66 | |
| GenBank mRNA | AK004148.1 | - | 5 - 6 | 819 | 66 | |
| BC089625.1 | - | 5 - 6 | 806 | 6756382 | 66 | |
| BC132636.1 | - | 5 - 6 | 807 | 40130913 | 66 | |
| BC132638.1 | - | 5 - 6 | 807 | 40130915 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mybphl
Genomic Map