Assay Details
| Gene Symbol: Stxbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011504.1 | NP_035634.1 | 13 - 14 | 1138 | 120 | |
| GenBank mRNA | AK138609.1 | - | 13 - 14 | 1170 | 120 | |
| AK151466.1 | - | 13 - 14 | 1133 | 120 | ||
| AK152261.1 | - | 13 - 14 | 1127 | 120 | ||
| AK155133.1 | - | 13 - 14 | 1136 | 120 | ||
| BC062901.1 | - | 13 - 14 | 1147 | 6810254 | 120 | |
| BC094055.1 | - | 13 - 14 | 1146 | 120 | ||
| D30798.1 | - | 13 - 14 | 1138 | 120 | ||
| U19521.1 | - | 13 - 14 | 1263 | 120 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stxbp3
Genomic Map