Assay Details
| Gene Symbol: Ampd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289719.1 | NP_001276648.1 | 11 - 12 | 1548 | 114 | |
| NM_001289720.1 | NP_001276649.1 | 10 - 11 | 1365 | 114 | ||
| NM_028779.5 | NP_083055.1 | 10 - 11 | 1634 | 114 | ||
| GenBank mRNA | AK004759.2 | - | 11 - 12 | 1533 | 114 | |
| AK034844.1 | - | 10 - 11 | 1338 | 115 | ||
| AK169980.1 | - | 11 - 12 | 1534 | 114 | ||
| BC049119.1 | - | 10 - 11 | 1595 | 6812571 | 114 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ampd2
Genomic Map