Assay Details
| Gene Symbol: Ampd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289719.1 | NP_001276648.1 | 16 - 17 | 2259 | 80 | |
| NM_001289720.1 | NP_001276649.1 | 15 - 16 | 2076 | 80 | ||
| NM_028779.5 | NP_083055.1 | 15 - 16 | 2345 | 80 | ||
| GenBank mRNA | AK004759.2 | - | 16 - 17 | 2244 | 80 | |
| AK034844.1 | - | 15 - 16 | 2050 | 80 | ||
| AK169980.1 | - | 16 - 17 | 2245 | 80 | ||
| BC016662.1 | - | 4 - 5 | 569 | 80 | ||
| BC049119.1 | - | 15 - 16 | 2306 | 6812571 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ampd2
Genomic Map