Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300021389.mm.1 |
NM_001289719.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021389.mm.1 |
NM_001289720.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021389.mm.1 |
NM_028779.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021389.mm.1 |
TC0300002730.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300021392.mm.1 |
NM_001289719.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021392.mm.1 |
NM_001289720.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021392.mm.1 |
NM_028779.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300021392.mm.1 |
TC0300002730.mm.1 |
MTA TC |