Assay Details
| Gene Symbol: Sds | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145565.1 | NP_663540.1 | 3 - 4 | 243 | 129 | |
| GenBank mRNA | AK149438.1 | - | 3 - 4 | 260 | 129 | |
| BC011259.1 | - | 3 - 4 | 240 | 129 | ||
| BC021605.1 | - | 3 - 4 | 240 | 5097741 | 129 | |
| BC021950.1 | - | 3 - 4 | 243 | 5102037 | 129 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sds
Genomic Map