Assay Details
| Gene Symbol: Sds | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145565.1 | NP_663540.1 | 5 - 6 | 479 | 85 | |
| GenBank mRNA | BC011259.1 | - | 5 - 6 | 476 | 85 | |
| BC021605.1 | - | 5 - 6 | 476 | 5097741 | 85 | |
| BC021950.1 | - | 5 - 6 | 479 | 5102037 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sds
Genomic Map