Assay Details
| Gene Symbol: Rbm19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028762.1 | NP_083038.1 | 3 - 4 | 443 | 68 | |
| GenBank mRNA | AK053511.1 | - | 3 - 4 | 447 | 68 | |
| AK146493.1 | - | 3 - 4 | 473 | 68 | ||
| AK167100.1 | - | 3 - 4 | 439 | 68 | ||
| BC025619.1 | - | 3 - 4 | 443 | 5320947 | 68 | |
| BC034010.1 | - | 3 - 4 | 409 | 4225239 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rbm19
Genomic Map