Assay Details
| Gene Symbol: Rasal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001281999.1 | NP_001268928.1 | 6 - 7 | 792 | 73 | |
| NM_013832.4 | NP_038860.2 | 7 - 8 | 763 | 73 | ||
| GenBank mRNA | AF086714.1 | - | 6 - 7 | 635 | 73 | |
| AK156050.1 | - | 6 - 7 | 756 | 73 | ||
| BC005418.1 | - | 7 - 8 | 777 | 3488024 | 73 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001281999.1 transcripts.
Set Membership: |
|

Rasal1
Genomic Map