Assay Details
| Gene Symbol: Stx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286033.1 | NP_001272962.1 | 5 - 6 | 507 | 62 | |
| NM_001286034.1 | NP_001272963.1 | 5 - 6 | 502 | 62 | ||
| NM_007941.3 | NP_031967.2 | 5 - 6 | 507 | 62 | ||
| GenBank mRNA | AK146462.1 | - | 5 - 6 | 436 | 62 | |
| AK165552.1 | - | 5 - 6 | 435 | 62 | ||
| AK167577.1 | - | 5 - 6 | 470 | 62 | ||
| AK168067.1 | - | 5 - 6 | 485 | 62 | ||
| BC046279.1 | - | 5 - 6 | 431 | 6392250 | 62 | |
| D10475.1 | - | 5 - 6 | 510 | 62 | ||
More Information
Set Membership: |
|

Stx2
Genomic Map