Assay Details
| Gene Symbol: Plbd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023625.4 | NP_076114.2 | 4 - 5 | 690 | 66 | |
| GenBank mRNA | AF217003.1 | - | 4 - 5 | 671 | 66 | |
| AK029813.1 | - | 4 - 5 | 690 | 66 | ||
| AK030234.1 | - | 4 - 5 | 642 | 66 | ||
| AK032930.1 | - | 4 - 5 | 686 | 66 | ||
| AK042089.1 | - | 686 | 66 | |||
| AK043608.1 | - | 4 - 5 | 680 | 66 | ||
| AK046937.1 | - | 4 - 5 | 680 | 66 | ||
| AK150201.1 | - | 4 - 5 | 681 | 66 | ||
| AK164220.1 | - | 4 - 5 | 671 | 66 | ||
| AK170387.1 | - | 4 - 5 | 682 | 66 | ||
| AK170631.1 | - | 4 - 5 | 670 | 66 | ||
| AK194379.1 | - | 1 - 2 | 66 | |||
| BC026395.1 | - | 4 - 5 | 668 | 66 | ||
| BC038605.1 | - | 4 - 5 | 671 | 4921245 | 66 | |
| BC048826.1 | - | 4 - 5 | 677 | 4196604 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Plbd2
Genomic Map