Assay Details
| Gene Symbol: Plbd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023625.4 | NP_076114.2 | 5 - 6 | 903 | 77 | |
| GenBank mRNA | AF217003.1 | - | 5 - 6 | 884 | 77 | |
| AK029813.1 | - | 5 - 6 | 903 | 77 | ||
| AK030234.1 | - | 5 - 6 | 855 | 77 | ||
| AK032930.1 | - | 5 - 6 | 899 | 77 | ||
| AK042089.1 | - | 899 | 77 | |||
| AK043608.1 | - | 5 - 6 | 893 | 77 | ||
| AK046937.1 | - | 5 - 6 | 893 | 77 | ||
| AK150201.1 | - | 5 - 6 | 892 | 77 | ||
| AK164220.1 | - | 5 - 6 | 884 | 77 | ||
| AK170387.1 | - | 5 - 6 | 895 | 77 | ||
| AK170631.1 | - | 5 - 6 | 883 | 77 | ||
| BC026395.1 | - | 5 - 6 | 881 | 77 | ||
| BC038605.1 | - | 5 - 6 | 884 | 4921245 | 77 | |
| BC048826.1 | - | 5 - 6 | 890 | 4196604 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Plbd2
Genomic Map