Assay Details
| Gene Symbol: Plbd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023625.4 | NP_076114.2 | 6 - 7 | 1002 | 88 | |
| GenBank mRNA | AF217003.1 | - | 6 - 7 | 983 | 88 | |
| AK004973.1 | - | 6 - 7 | 1005 | 88 | ||
| AK029813.1 | - | 6 - 7 | 1002 | 88 | ||
| AK030234.1 | - | 6 - 7 | 954 | 88 | ||
| AK032930.1 | - | 6 - 7 | 998 | 88 | ||
| AK042089.1 | - | 998 | 88 | |||
| AK043608.1 | - | 6 - 7 | 992 | 88 | ||
| AK046937.1 | - | 6 - 7 | 992 | 88 | ||
| AK150201.1 | - | 6 - 7 | 991 | 88 | ||
| AK164220.1 | - | 6 - 7 | 983 | 88 | ||
| AK170387.1 | - | 6 - 7 | 994 | 88 | ||
| AK170631.1 | - | 6 - 7 | 982 | 88 | ||
| BC026395.1 | - | 6 - 7 | 980 | 88 | ||
| BC038605.1 | - | 6 - 7 | 983 | 4921245 | 88 | |
| BC048826.1 | - | 6 - 7 | 989 | 4196604 | 88 | |
| BC060095.1 | - | 4 - 5 | 2593 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Plbd2
Genomic Map