Assay Details
| Gene Symbol: Anapc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019805.4 | NP_062779.3 | 6 - 7 | 956 | 75 | |
| GenBank mRNA | AK049968.1 | - | 6 - 7 | 895 | 75 | |
| AK137286.1 | - | 6 - 7 | 920 | 75 | ||
| AK150764.1 | - | 6 - 7 | 920 | 75 | ||
| AK150891.1 | - | 6 - 7 | 920 | 75 | ||
| AK153114.1 | - | 6 - 7 | 920 | 75 | ||
| AK159523.1 | - | 6 - 7 | 888 | 75 | ||
| AK164202.1 | - | 6 - 7 | 925 | 75 | ||
| BC006635.1 | - | 6 - 7 | 909 | 3482261 | 75 | |
More Information
Set Membership: |
|

Anapc7
Genomic Map