Assay Details
| Gene Symbol: Anapc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019805.4 | NP_062779.3 | 8 - 9 | 1259 | 97 | |
| GenBank mRNA | AF076607.2 | - | 8 - 9 | 1223 | 97 | |
| AK030775.1 | - | 8 - 9 | 1224 | 97 | ||
| AK049968.1 | - | 8 - 9 | 1198 | 97 | ||
| AK137286.1 | - | 8 - 9 | 1223 | 97 | ||
| AK150764.1 | - | 8 - 9 | 1223 | 97 | ||
| AK150891.1 | - | 8 - 9 | 1223 | 97 | ||
| AK153114.1 | - | 8 - 9 | 1223 | 97 | ||
| AK159523.1 | - | 8 - 9 | 1191 | 97 | ||
| AK164202.1 | - | 8 - 9 | 1228 | 97 | ||
| BC006635.1 | - | 8 - 9 | 1212 | 3482261 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Anapc7
Genomic Map