Assay Details
| Gene Symbol: Arpc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019824.3 | NP_062798.1 | 1 - 2 | 69 | 71 | |
| GenBank mRNA | AK003485.1 | - | 1 - 2 | 100 | 71 | |
| AK005437.1 | - | 1 - 2 | 88 | 71 | ||
| AK171651.1 | - | 1 - 2 | 84 | 71 | ||
| BC013618.1 | - | 1 - 2 | 69 | 4013537 | 71 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK003485.1, AK005437.1, AK171651.1, BC013618.1, NM_019824.3 transcripts.
Set Membership: |
|

Arpc3
Genomic Map