Assay Details
| Gene Symbol: Arpc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019824.3 | NP_062798.1 | 3 - 4 | 244 | 128 | |
| GenBank mRNA | AB038243.1 | - | 2 - 3 | 180 | 128 | |
| AK003485.1 | - | 3 - 4 | 275 | 128 | ||
| AK005437.1 | - | 3 - 4 | 263 | 128 | ||
| AK171651.1 | - | 3 - 4 | 259 | 128 | ||
| BC013618.1 | - | 3 - 4 | 244 | 4013537 | 128 | |
| BC054440.1 | - | 3 - 4 | 211 | 6395280 | 128 | |
| BC055036.1 | - | 3 - 4 | 211 | 128 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arpc3
Genomic Map