| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500012051.mm.1 |
TC0500001370.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500012051.mm.1 |
NM_009125.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500012051.mm.1 |
NM_009125.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500012048.mm.1 |
TC0500001370.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500012048.mm.1 |
NM_009125.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500012048.mm.1 |
NM_009125.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500012050.mm.1 |
TC0500001370.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500012050.mm.1 |
NM_009125.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500012050.mm.1 |
NM_009125.2 |
Refseq |