Assay Details
| Gene Symbol: Atxn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009125.2 | NP_033151.2 | 10 - 11 | 1788 | 68 | |
| GenBank mRNA | AF041472.1 | - | 10 - 11 | 1787 | 68 | |
| AK135976.1 | - | 5 - 6 | 745 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atxn2
Genomic Map