Assay Details
| Gene Symbol: Rps5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009095.2 | NP_033121.2 | 4 - 5 | 528 | 99 | |
| GenBank mRNA | AK002235.1 | - | 4 - 5 | 522 | 99 | |
| AK009663.1 | - | 4 - 5 | 522 | 99 | ||
| AK010681.1 | - | 4 - 5 | 522 | 99 | ||
| AK012424.1 | - | 4 - 5 | 522 | 99 | ||
| AK012481.1 | - | 4 - 5 | 522 | 99 | ||
| AK020477.1 | - | 4 - 5 | 521 | 99 | ||
| AK020754.1 | - | 4 - 5 | 522 | 99 | ||
| AK050611.1 | - | 4 - 5 | 522 | 99 | ||
| AK050622.1 | - | 4 - 5 | 522 | 99 | ||
| AK152449.1 | - | 4 - 5 | 522 | 99 | ||
| AK160552.1 | - | 4 - 5 | 522 | 99 | ||
| AK164079.1 | - | 4 - 5 | 522 | 99 | ||
| AK167533.1 | - | 4 - 5 | 522 | 99 | ||
| AK168044.1 | - | 4 - 5 | 522 | 99 | ||
| AK168106.1 | - | 4 - 5 | 522 | 99 | ||
| AK169016.1 | - | 4 - 5 | 522 | 99 | ||
| AK172446.1 | - | 4 - 5 | 522 | 99 | ||
| BC020401.1 | - | 4 - 5 | 499 | 99 | ||
| BC058690.1 | - | 4 - 5 | 509 | 6814814 | 99 | |
| Y12431.1 | - | 4 - 5 | 504 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24973639 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24973637 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24973634 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24973640 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24973638 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24973635 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2063992 | Y12431.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2063992 | BC020401.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2063992 | BC058690.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2063992 | U78085.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2063992 | AK020477.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 204839 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 253936 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 928585 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P171292 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2063992 | Y12431.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2063992 | BC020401.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2063992 | BC058690.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2063992 | U78085.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2063992 | AK020477.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P174335 | Y12431.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P174335 | AK167533.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P174335 | AK168044.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P174335 | AK168106.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P174335 | AK169016.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24973661 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24973649 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24973617 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24973638 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24973650 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24973647 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24973633 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24973622 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24973664 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24973652 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24973629 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24973644 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24973621 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24973627 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24973624 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24973636 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24973665 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24973628 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24973657 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24973663 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24973637 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24973625 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24973640 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24973660 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24973623 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24973670 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24973631 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24973634 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24973653 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24973619 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24973668 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24973642 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P171292 | - | - |
| Whole Mouse Genome G4122A | A_51_P174335 | Y12431.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P174335 | AK167533.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P174335 | AK168044.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P174335 | AK168106.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P174335 | AK169016.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134680 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P171292 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05972809 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05972807 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05972795 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06452168 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05972804 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05972802 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05972800 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24973638 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24973635 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24973639 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24973637 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24973640 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24973634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973647 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05972804 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24973623 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06322301 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24973642 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06296677 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05972795 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06296678 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24973633 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24973621 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24973658 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24973665 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06335688 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06335696 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06296672 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05972811 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24973654 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06479309 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24973653 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06296673 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06649065 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24973617 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24973636 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05972798 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24973631 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05904292 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24973670 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24973625 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06479310 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06548164 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24973619 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05964811 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24973647 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24973650 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24973668 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06572852 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05972801 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24973612 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24973628 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10550056 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550053 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550057 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550058 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550055 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550054 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416054_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10550052 | - | - |
| Mouse Genome 430 2.0 Array | 1416054_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1416054_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20103_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20103_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700001157.mm.1 | TC0700000151.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700001157.mm.1 | NM_009095.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700001157.mm.1 | NM_009095.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700001153.mm.1 | TC0700000151.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700001153.mm.1 | NM_009095.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700001153.mm.1 | NM_009095.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700001158.mm.1 | TC0700000151.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700001158.mm.1 | NM_009095.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700001158.mm.1 | NM_009095.2 | Refseq |
| Mouse Expression Set 430 | 1416054_PM_at | - | - |
| Mouse Expression Set 430 | 1416054_at | - | - |
| Murine Genome U74v2 Set | 99336_PM_at | - | - |
| Murine Genome U74v2 Set | 99336_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416054_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416054_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416054_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416054_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 580093 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000580093 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1236158 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1236158 | - | - |
| mouse-6 v1.1 (Illumina) | scl33074.8.142_21-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 580093 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl33074.8.142_21-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 580093 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2808946 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1236158 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2808946 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl33074.8.142_21-S | - | - |

Rps5
Genomic Map