Assay Details
Gene Symbol: Sult2a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199306.1 | NP_001186235.1 | 4 - 5 | 692 | 88 | |
NM_175250.5 | NP_780459.1 | 4 - 5 | 742 | 88 | ||
GenBank mRNA | AK041070.1 | - | 4 - 5 | 742 | 88 | |
AK050422.1 | - | 4 - 5 | 690 | 88 | ||
BC031851.1 | - | 4 - 5 | 638 | 4193548 | 88 | |
EU486166.1 | - | 4 - 5 | 742 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
