Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700021090.mm.1 |
TC0700002383.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700021090.mm.1 |
NM_001199306.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021090.mm.1 |
NM_001199306.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021091.mm.1 |
TC0700002383.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700021091.mm.1 |
NM_175250.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021091.mm.1 |
NM_175250.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021087.mm.1 |
TC0700002383.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700021087.mm.1 |
NM_001199306.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021087.mm.1 |
NM_001199306.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021087.mm.1 |
NM_175250.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021087.mm.1 |
NM_175250.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021089.mm.1 |
TC0700002383.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700021089.mm.1 |
NM_001199306.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021089.mm.1 |
NM_001199306.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021085.mm.1 |
TC0700002383.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700021085.mm.1 |
NM_001199306.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021085.mm.1 |
NM_001199306.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021085.mm.1 |
NM_175250.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021085.mm.1 |
NM_175250.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021086.mm.1 |
TC0700002383.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700021086.mm.1 |
NM_001199306.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021086.mm.1 |
NM_001199306.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021086.mm.1 |
NM_175250.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021086.mm.1 |
NM_175250.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021084.mm.1 |
TC0700002383.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700021084.mm.1 |
NM_001199306.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021084.mm.1 |
NM_001199306.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021084.mm.1 |
NM_175250.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021084.mm.1 |
NM_175250.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021088.mm.1 |
TC0700002383.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700021088.mm.1 |
NM_001199306.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021088.mm.1 |
NM_001199306.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021088.mm.1 |
NM_175250.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021088.mm.1 |
NM_175250.5 |
Refseq |