Assay Details
| Gene Symbol: Ahcyl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145542.3 | NP_663517.2 | 14 - 15 | 1661 | 96 | |
| GenBank mRNA | AB092504.1 | - | 14 - 15 | 1390 | 96 | |
| AK078823.1 | - | 7 - 8 | 863 | 96 | ||
| BC018218.1 | - | 14 - 15 | 1490 | 4007102 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ahcyl1
Genomic Map