Assay Details
| Gene Symbol: Slc6a17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293689.1 | NP_001280618.1 | 3 - 4 | 946 | 108 | |
| NM_172271.2 | NP_758475.1 | 3 - 4 | 946 | 108 | ||
| NR_121629.1 | 2 - 3 | 533 | 108 | |||
| GenBank mRNA | AK032262.1 | - | 2 - 3 | 323 | 108 | |
| AK083807.1 | - | 3 - 4 | 847 | 108 | ||
| AK085620.1 | - | 3 - 4 | 680 | 109 | ||
| AK163113.1 | - | 3 - 4 | 950 | 108 | ||
| AY155578.1 | - | 3 - 4 | 947 | 108 | ||
| BC033610.1 | - | 3 - 4 | 571 | 108 | ||
| BC141148.1 | - | 2 - 3 | 517 | 9055760 | 108 | |
| BC171962.1 | - | 2 - 3 | 517 | 9054248 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc6a17
Genomic Map