Assay Details
| Gene Symbol: Dram2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286986.1 | NP_001273915.1 | 7 - 8 | 857 | 148 | |
| NM_001286987.1 | NP_001273916.1 | 6 - 7 | 735 | 148 | ||
| NM_026013.3 | NP_080289.1 | 7 - 8 | 860 | 148 | ||
| GenBank mRNA | AK009940.1 | - | 7 - 8 | 797 | 148 | |
| AK012044.1 | - | 7 - 8 | 777 | 148 | ||
| AK015888.1 | - | 7 - 8 | 823 | 148 | ||
| AK052824.1 | - | 6 - 7 | 669 | 148 | ||
| AK172455.1 | - | 7 - 8 | 827 | 148 | ||
More Information
Set Membership: |
|

Dram2
Genomic Map