Assay Details
| Gene Symbol: Itgbl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145467.2 | NP_663442.2 | 5 - 6 | 895 | 85 | |
| GenBank mRNA | AK029068.1 | - | 5 - 6 | 862 | 85 | |
| BC020152.1 | - | 5 - 6 | 862 | 3708340 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Itgbl1
Genomic Map