Assay Details
| Gene Symbol: Magohb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025564.2 | NP_079840.2 | 3 - 4 | 342 | 147 | |
| GenBank mRNA | AK006303.1 | - | 3 - 4 | 332 | 147 | |
| AK007769.1 | - | 3 - 4 | 329 | 147 | ||
| AK008200.1 | - | 3 - 4 | 344 | 147 | ||
| AK013310.1 | - | 3 - 4 | 327 | 147 | ||
| BC027505.1 | - | 3 - 4 | 315 | 3372083 | 147 | |
| KF051005.1 | - | 3 - 4 | 263 | 147 | ||
More Information
Set Membership: |
|

Magohb
Genomic Map