Assay Details
| Gene Symbol: Ptcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133735.2 | NP_598496.3 | 6 - 7 | 1790 | 94 | |
| GenBank mRNA | AK088764.1 | - | 6 - 7 | 1709 | 94 | |
| AK143877.1 | - | 6 - 7 | 1748 | 94 | ||
| AK150514.1 | - | 6 - 7 | 1747 | 94 | ||
| AK151064.1 | - | 6 - 7 | 1758 | 94 | ||
| AK152077.1 | - | 6 - 7 | 1758 | 94 | ||
| AK153366.1 | - | 6 - 7 | 1758 | 94 | ||
| AK171484.1 | - | 6 - 7 | 1745 | 94 | ||
| BC004766.1 | - | 6 - 7 | 1708 | 3156304 | 94 | |
| BC019121.1 | - | 4 - 5 | 1178 | 94 | ||
| BC083097.1 | - | 6 - 7 | 1749 | 30459747 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ptcd1
Genomic Map