Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700013696.mm.1 |
NM_001199048.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013696.mm.1 |
TC1700001563.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700013698.mm.1 |
NM_001199048.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013698.mm.1 |
TC1700001563.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700013699.mm.1 |
NM_001199048.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013699.mm.1 |
TC1700001563.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700013700.mm.1 |
NM_001199048.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013700.mm.1 |
TC1700001563.mm.1 |
MTA TC |