Assay Details
| Gene Symbol: Gpaa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010331.2 | NP_034461.1 | 1 - 2 | 177 | 73 | |
| GenBank mRNA | AB002136.1 | - | 1 - 2 | 106 | 73 | |
| AB006970.1 | - | 1 - 2 | 113 | 73 | ||
| AK077518.1 | - | 1 - 2 | 177 | 73 | ||
| AK164219.1 | - | 1 - 2 | 123 | 73 | ||
| AK164978.1 | - | 1 - 2 | 177 | 73 | ||
| BC006697.1 | - | 1 - 2 | 116 | 3498573 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpaa1
Genomic Map