Assay Details
| Gene Symbol: Gpaa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010331.2 | NP_034461.1 | 5 - 6 | 712 | 89 | |
| GenBank mRNA | AB002136.1 | - | 5 - 6 | 641 | 89 | |
| AB006970.1 | - | 5 - 6 | 648 | 89 | ||
| AK077518.1 | - | 5 - 6 | 712 | 89 | ||
| AK164219.1 | - | 5 - 6 | 658 | 89 | ||
| AK164978.1 | - | 5 - 6 | 712 | 89 | ||
| AK168057.1 | - | 5 - 6 | 915 | 89 | ||
| BC006697.1 | - | 5 - 6 | 651 | 3498573 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpaa1
Genomic Map