Assay Details
| Gene Symbol: Pycrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025412.2 | NP_079688.2 | 1 - 2 | 189 | 55 | |
| GenBank mRNA | AK002912.1 | - | 1 - 2 | 120 | 55 | |
| AK004291.1 | - | 1 - 2 | 193 | 55 | ||
| AK013214.1 | - | 1 - 2 | 115 | 55 | ||
| BC026536.1 | - | 1 - 2 | 110 | 4987954 | 55 | |
| CX212991.1 | - | 1 - 2 | 196 | 55 | ||
More Information
Set Membership: |
|

Pycrl
Genomic Map