Assay Details
| Gene Symbol: Pycrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025412.2 | NP_079688.2 | 3 - 4 | 429 | 69 | |
| GenBank mRNA | AK002912.1 | - | 3 - 4 | 360 | 69 | |
| AK013214.1 | - | 3 - 4 | 355 | 69 | ||
| BC026536.1 | - | 3 - 4 | 350 | 4987954 | 69 | |
| CX212991.1 | - | 3 - 4 | 436 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pycrl
Genomic Map