| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1500014000.mm.1 |
NM_001110827.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014000.mm.1 |
NM_001110828.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014000.mm.1 |
NM_001110829.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014000.mm.1 |
NM_001110830.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014000.mm.1 |
NM_001286417.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014000.mm.1 |
NM_001286418.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014000.mm.1 |
NM_001286419.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014000.mm.1 |
NM_053104.6 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014000.mm.1 |
NM_175387.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014000.mm.1 |
NM_175387.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500014000.mm.1 |
TC1500001799.mm.1 |
MTA TC |