Assay Details
| Gene Symbol: Mtus1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005863.2 | NP_001005863.1 | 10 - 11 | 3519 | 77 | |
| NM_001005864.3 | NP_001005864.1 | 8 - 9 | 1125 | 77 | ||
| NM_001005865.3 | NP_001005865.2 | 6 - 7 | 1138 | 77 | ||
| NM_001286413.1 | NP_001273342.1 | 10 - 11 | 3401 | 77 | ||
| GenBank mRNA | AF173380.1 | - | 6 - 7 | 977 | 77 | |
| AF493235.1 | - | 6 - 7 | 1042 | 77 | ||
| AK030510.1 | - | 6 - 7 | 1102 | 77 | ||
| AK031693.1 | - | 6 - 7 | 1072 | 77 | ||
| AK035576.1 | - | 4 - 5 | 506 | 77 | ||
| AK129324.1 | - | 4 - 5 | 3615 | 77 | ||
| AK149529.1 | - | 1 - 2 | 565 | 77 | ||
| AY246699.1 | - | 6 - 7 | 800 | 77 | ||
| AY626781.1 | - | 6 - 7 | 1135 | 77 | ||
| AY626782.1 | - | 8 - 9 | 1123 | 77 | ||
| AY626783.1 | - | 9 - 10 | 3341 | 77 | ||
| BC041777.1 | - | 9 - 10 | 2184 | 4240274 | 77 | |
| BC042206.1 | - | 9 - 10 | 2184 | 4240274 | 77 | |
| BC043321.1 | - | 8 - 9 | 1089 | 77 | ||
| BC089009.1 | - | 10 - 11 | 3401 | 30543240 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mtus1
Genomic Map