Assay Details
| Gene Symbol: Vps37a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033560.3 | NP_291038.2 | 10 - 11 | 1442 | 114 | |
| GenBank mRNA | AK016385.1 | - | 10 - 11 | 1425 | 114 | |
| AK157507.1 | - | 10 - 11 | 1409 | 114 | ||
| AK171055.1 | - | 10 - 11 | 1431 | 114 | ||
| BC039290.1 | - | 10 - 11 | 1398 | 5066508 | 114 | |
More Information
Updated Mapping Information
Set Membership: |
|

Vps37a
Genomic Map