Assay Details
| Gene Symbol: Vps37a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033560.3 | NP_291038.2 | 3 - 4 | 637 | 150 | |
| GenBank mRNA | AK048942.1 | - | 3 - 4 | 617 | 150 | |
| AK054055.1 | - | 3 - 4 | 619 | 150 | ||
| AK137479.1 | - | 3 - 4 | 622 | 150 | ||
| AK157507.1 | - | 3 - 4 | 603 | 150 | ||
| AK170280.1 | - | 3 - 4 | 606 | 150 | ||
| AK170943.1 | - | 3 - 4 | 595 | 150 | ||
| AK170961.1 | - | 3 - 4 | 626 | 150 | ||
| AK171055.1 | - | 3 - 4 | 626 | 150 | ||
| AK171221.1 | - | 3 - 4 | 638 | 150 | ||
| BC039290.1 | - | 3 - 4 | 593 | 5066508 | 150 | |
| BC110396.1 | - | 3 - 4 | 552 | 5253683 | 150 | |
More Information
Updated Mapping Information
Set Membership: |
|

Vps37a
Genomic Map