Assay Details
| Gene Symbol: Vps37a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033560.3 | NP_291038.2 | 6 - 7 | 1038 | 72 | |
| GenBank mRNA | AK016385.1 | - | 6 - 7 | 1021 | 72 | |
| AK048942.1 | - | 6 - 7 | 1018 | 72 | ||
| AK054055.1 | - | 6 - 7 | 1020 | 72 | ||
| AK137479.1 | - | 6 - 7 | 1023 | 72 | ||
| AK157507.1 | - | 6 - 7 | 1005 | 72 | ||
| AK170280.1 | - | 6 - 7 | 1007 | 72 | ||
| AK170943.1 | - | 6 - 7 | 996 | 72 | ||
| AK170961.1 | - | 6 - 7 | 1027 | 72 | ||
| AK171055.1 | - | 6 - 7 | 1027 | 72 | ||
| AK171221.1 | - | 6 - 7 | 1039 | 72 | ||
| AK218075.1 | - | 1 - 2 | 39 | 72 | ||
| BC039290.1 | - | 6 - 7 | 994 | 5066508 | 72 | |
| BC110396.1 | - | 6 - 7 | 953 | 5253683 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Vps37a
Genomic Map