Assay Details
| Gene Symbol: Vps37a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033560.3 | NP_291038.2 | 7 - 8 | 1160 | 115 | |
| GenBank mRNA | AK016385.1 | - | 7 - 8 | 1143 | 115 | |
| AK054055.1 | - | 7 - 8 | 1142 | 115 | ||
| AK137479.1 | - | 7 - 8 | 1145 | 115 | ||
| AK157507.1 | - | 7 - 8 | 1127 | 115 | ||
| AK170280.1 | - | 7 - 8 | 1129 | 115 | ||
| AK170943.1 | - | 7 - 8 | 1118 | 115 | ||
| AK170961.1 | - | 7 - 8 | 1149 | 115 | ||
| AK171055.1 | - | 7 - 8 | 1149 | 115 | ||
| AK171221.1 | - | 7 - 8 | 1161 | 115 | ||
| BC039290.1 | - | 7 - 8 | 1116 | 5066508 | 115 | |
| BC110396.1 | - | 7 - 8 | 1075 | 5253683 | 115 | |
More Information
Updated Mapping Information
Set Membership: |
|

Vps37a
Genomic Map