Assay Details
| Gene Symbol: Vps37a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033560.3 | NP_291038.2 | 8 - 9 | 1223 | 96 | |
| GenBank mRNA | AK016385.1 | - | 8 - 9 | 1206 | 96 | |
| AK048942.1 | - | 7 - 8 | 1285 | 96 | ||
| AK157507.1 | - | 8 - 9 | 1190 | 96 | ||
| AK170280.1 | - | 8 - 9 | 1192 | 96 | ||
| AK170943.1 | - | 8 - 9 | 1181 | 96 | ||
| AK170961.1 | - | 8 - 9 | 1212 | 96 | ||
| AK171055.1 | - | 8 - 9 | 1212 | 96 | ||
| AK171221.1 | - | 8 - 9 | 1224 | 96 | ||
| AK218075.1 | - | 3 - 4 | 224 | 96 | ||
| BC039290.1 | - | 8 - 9 | 1179 | 5066508 | 96 | |
| BC110396.1 | - | 8 - 9 | 1138 | 5253683 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Vps37a
Genomic Map