Assay Details
| Gene Symbol: Ankrd37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039562.1 | NP_001034651.1 | 3 - 4 | 482 | 60 | |
| GenBank mRNA | AK020869.1 | - | 3 - 4 | 466 | 60 | |
| AK075868.1 | - | 3 - 4 | 356 | 60 | ||
| BC039642.1 | - | 3 - 4 | 480 | 5352548 | 60 | |
| BC092377.1 | - | 3 - 4 | 482 | 6818571 | 60 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ankrd37
Genomic Map