Assay Details
| Gene Symbol: Stx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152220.2 | NP_689344.1 | 10 - 11 | 1573 | 67 | |
| GenBank mRNA | AK133830.1 | - | 10 - 11 | 1096 | 67 | |
| AK171135.1 | - | 10 - 11 | 1116 | 67 | ||
| AW988213.1 | - | 4 - 5 | 410 | 67 | ||
| D29797.1 | - | 10 - 11 | 994 | 67 | ||
More Information
Set Membership: |
|

Stx3
Genomic Map