Assay Details
| Gene Symbol: Stx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152220.2 | NP_689344.1 | 8 - 9 | 1356 | 98 | |
| GenBank mRNA | AK133830.1 | - | 8 - 9 | 879 | 98 | |
| AK170105.1 | - | 8 - 9 | 766 | 99 | ||
| AK171135.1 | - | 8 - 9 | 899 | 98 | ||
| AW988213.1 | - | 2 - 3 | 193 | 98 | ||
| D29797.1 | - | 8 - 9 | 777 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stx3
Genomic Map