Assay Details
| Gene Symbol: Tmem216 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277860.1 | NP_001264789.1 | 4 - 5 | 447 | 76 | |
| NM_001277861.1 | NP_001264790.1 | 4 - 5 | 461 | 76 | ||
| NM_026798.3 | NP_081074.1 | 4 - 5 | 520 | 76 | ||
| GenBank mRNA | AF332866.1 | - | 3 - 4 | 386 | 76 | |
| AK003740.1 | - | 4 - 5 | 522 | 76 | ||
| AK013286.1 | - | 4 - 5 | 451 | 76 | ||
| BC052053.1 | - | 4 - 5 | 424 | 5693205 | 76 | |
| BY704000.1 | - | 4 - 5 | 522 | 76 | ||
| DV063021.1 | - | 4 - 5 | 515 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31951353 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31951351 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31951354 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2075268 | AK013286.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2075268 | BC052053.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2075268 | NM_026798.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2075268 | AK003740.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1993719 | BC052053.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1993719 | NM_026798.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1993719 | AK003740.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 689915 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 773169 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 695721 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 130173 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P136023 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1993719 | BC052053.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1993719 | NM_026798.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1993719 | AK003740.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2075268 | AK013286.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2075268 | BC052053.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2075268 | NM_026798.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2075268 | AK003740.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31951371 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31951352 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31951363 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31951356 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31951364 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31951341 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31951366 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31951374 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31951334 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31951355 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31951370 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31951353 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31951362 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31951360 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31951347 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31951349 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31951342 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31951368 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31951343 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31951373 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31951372 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31951339 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31951358 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31951336 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31951345 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03549186 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P136023 | - | - |
| Whole Mouse Genome G4122A | A_52_P619903 | AK013286.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P619903 | BC052053.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P619903 | NM_026798.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P619903 | AF332866.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P619903 | AK003740.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P103911 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P136023 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14203229 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14203224 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14203225 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14203226 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14203223 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14395009 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14395006 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14395008 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14203220 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31951353 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31951354 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31951351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951385 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951386 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951360 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951320 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951385 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14177896 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951336 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14418095 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14383573 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14205529 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951377 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14383574 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14383579 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951382 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14203226 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951318 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951358 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951372 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951387 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951356 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951326 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14318521 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951361 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951345 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14395009 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951371 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951343 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951341 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951368 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14318528 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951331 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14193862 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951334 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14203224 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951369 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951364 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951362 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951333 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951329 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14318523 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951383 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951310 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951366 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951339 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951349 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951374 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951315 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951347 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951373 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951367 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14203223 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951363 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10465994 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465993 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465997 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465996 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465995 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465999 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465998 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453567_PM_s_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1426217_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10465990 | - | - |
| Mouse Genome 430 2.0 Array | 1453567_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1426217_at | - | - |
| Mouse Genome 430 2.0 Array | 1453567_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1426217_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68642_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68642_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900010351.mm.1 | NM_001277860.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900010351.mm.1 | NM_001277861.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900010351.mm.1 | NM_026798.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900010351.mm.1 | TC1900001105.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900010349.mm.1 | NM_001277860.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900010349.mm.1 | NM_001277861.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900010349.mm.1 | NM_026798.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900010349.mm.1 | TC1900001105.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900010350.mm.1 | NM_001277860.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900010350.mm.1 | NM_001277861.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900010350.mm.1 | NM_026798.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900010350.mm.1 | TC1900001105.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1453567_s_at | - | - |
| Mouse Expression Set 430 | 1426217_PM_at | - | - |
| Mouse Expression Set 430 | 1453567_PM_s_at | - | - |
| Mouse Expression Set 430 | 1426217_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426217_at | - | - |
| Mouse Genome 430A 2.0 Array | 1453567_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426217_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1453567_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1453567_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1453567_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426217_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426217_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 914732 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4920427 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004920427 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1214321 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1214321 | - | - |
| mouse-6 v1.1 (Illumina) | scl068642.1_282-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4920427 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl068642.1_282-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4920427 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2799217 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2799217 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl068642.1_282-S | - | - |

Tmem216
Genomic Map