Assay Details
| Gene Symbol: Pms2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008886.2 | NP_032912.2 | 6 - 7 | 764 | 67 | |
| GenBank mRNA | AK034866.1 | - | 6 - 7 | 740 | 67 | |
| AK083049.1 | - | 6 - 7 | 721 | 67 | ||
| AK146365.1 | - | 6 - 7 | 777 | 67 | ||
| BC022179.1 | - | 6 - 7 | 732 | 67 | ||
| BC058189.1 | - | 6 - 7 | 772 | 67 | ||
| BC066110.1 | - | 6 - 7 | 764 | 67 | ||
| BC083502.1 | - | 6 - 7 | 724 | 67 | ||
| BC086637.1 | - | 6 - 7 | 740 | 67 | ||
| BC110684.1 | - | 6 - 7 | 717 | 67 | ||
| BC139774.1 | - | 6 - 7 | 770 | 67 | ||
| BC141287.1 | - | 6 - 7 | 740 | 9055899 | 67 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC139774.1 transcripts.
Set Membership: |
|

Pms2
Genomic Map